Specifications
- Description:Repositionable Dry Mounts
- Size:8×10"
- Cat. No.:100488-822
- Supplier no.:77604
Specifications
About this item
RDM is a microthin, clear Mylar® film coated on both sides with an adhesive, specially formulated for photographic purposes.
- Made of pressure-sensitive mounting material
- No heat is needed
This "delayed-set" adhesive allows for repositioning of prints before providing a permanent bond.
The Mylar® base will not absorb moisture or change its characteristics through time; thereby it eliminates the common mounting problems that are experienced with other mounting materials.
RDM sheets are made 1/4" oversize which eliminates alignment problems and makes it easier to use.