Replacement Stopcock, Electron Microscopy Sciences
Product Details & Documents
A polypropylene body which has a TFE plug and washer as well as a bright red handle and nut.
Its taper fits on the desiccator port and the plug is 3-way allowing for vacuum draw, shut-off, and release. May be used with EMS vacuum desiccators.
Specifications
- Description:Replacement Stop Cock