Orders LinkContactUsLinkComponent
Inoculating Loop/Needle, Electron Microscopy Sciences
Inoculating Loop/Needle, Electron Microscopy Sciences
Inoculating Loop
Catalog # 100489-540
Supplier #:  72952-10
Inoculating Loop/Needle, Electron Microscopy Sciences
Catalog # 100489-540
Inoculating Loop/Needle, Electron Microscopy Sciences
Inoculating Loop/Needle, Electron Microscopy Sciences
Inoculating Loop
Price
$41.35
Log in for availability
Pack of 12
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Nichrome wire. An 8” aluminum handle with a 3” nichrome wire, 25 gauge. Loop O.D. is 4 mm; loop I.D. is 2.5 mm. Available with a loop or a needle.
Specifications
  • Description:
    Inoculating Loop