Indexed Grid Scales, Electron Microscopy Sciences
About this item
Useful for area of specimen determinations, especially rectangular shapes, also for particle counting.
3 Options Available Below
Product Details & Documents
Useful for area of specimen determinations, especially rectangular shapes, also for particle counting.
Product Family Options
Product Information
- SizeAvailabilityPrice
- 100499-164NE35 GraticuleSupplier Number: 68014-16
- 100499-166NE35 GraticuleSupplier Number: 68014-19
- 100499-168NE35 GraticuleSupplier Number: 68014-21