Orders LinkContactUsLinkComponent
Immersion Oils, Electron Microscopy Sciences
Immersion Oils, Electron Microscopy Sciences
Certificates
Immersion Oils, Electron Microscopy Sciences
Immersion Oils, Electron Microscopy Sciences
13 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Cargille PCB-Free Immersion oils were introduced in 1972. Today, no other immersion oil meets FDA criteria.  Now four standard types are available with the introduction of Type NVH, a very high viscosity oil. All Cargille Immersion Oils meet DIN 58 884 (Deutsche Institut Fur Normung e.V.).
For normal light mcroscopy: Types A and amp B, with viscosities of 150 and 1250 cSt respectively, meet these requirements. The greater the gap between cover glass and objective, or condenser and slide, the more desirable high viscosity becomes. For extremely large gaps, type NVH should be used. All cargille immersion ols can be blended for intermediate viscosities.
37 °C Immersion oils: A number of research projects require microscopic studies at internal body temperature.   Since the index/temperature relationship is inverse, increasing temperature significantly reduces the index of the immersion oil and consequently the imagery deteriorates.  This problem has been resolved by the formation of Cargille 37 °C Immersion Oils, which have an index of 1.515 at 37 °C. Consideration of these oils is also suggested where stage temperatures are elevated by sub-stage illuminators and high wattage projection equipment.
Immersion oil type 300: Automated hematology systems depend on accurate, precisely controlled physical and optical properties of immersion oil for successful imaging and mechanical processing. Type 300 is designed and manufactured to meet the stringent requirements of this equipment, which include specialized viscosity and exacting controls for its consistency.
High viscosity NVH: For horizontal, inverted and inclined instruments and projection equipment higher viscosities are generally required.  Type NVH meets the requirement, having a viscosity of 21000 cSt.
 : Certificates of analysis are available.
Product Family Options
Product Information
  • Size
    Availability
    Price
  • 100496-510
    Immersion Oil Type A
    Immersion Oil Type A
    1 oz.
    Log in for availability
    Log in for availability
    Each (1oz)
    Each (1oz) - $30.86
    $30.86
    Each (1oz)
       $30.86
    Supplier Number: 16907-01
  • 100496-512
    Immersion Oil Type A
    Immersion Oil Type A
    16 oz.
    Log in for availability
    Log in for availability
    Each (16oz)
    Each (16oz) - $130.94
    $130.94
    Each (16oz)
       $130.94
    Supplier Number: 16907-16
  • 100504-054
    Immersion Oil Type A
    Immersion Oil Type A
    120 mL
    Log in for availability
    Log in for availability
    Each (120ml)
    Each (120ml) - $63.36
    $63.36
    Each (120ml)
       $63.36
    Supplier Number: 16907
  • 100496-514
    Immersion Oil Type B
    Immersion Oil Type B
    1 oz.
    Log in for availability
    Log in for availability
    Each (1oz)
    Each (1oz) - $29.86
    $29.86
    Each (1oz)
       $29.86
    Supplier Number: 16908-01
  • 100496-516
    Immersion Oil Type B
    Immersion Oil Type B
    16 oz.
    Log in for availability
    Log in for availability
    Each (16oz)
    Each (16oz) - $130.94
    $130.94
    Each (16oz)
       $130.94
    Supplier Number: 16908-16
  • 100504-056
    Immersion Oil Type B
    Immersion Oil Type B
    120 mL
    Log in for availability
    Log in for availability
    Each (120ml)
    Each (120ml) - $63.32
    $63.32
    Each (120ml)
       $63.32
    Supplier Number: 16908
  • 100496-518
    Immersion Oil Type NVH
    Immersion Oil Type NVH
    1 oz.
    Log in for availability
    Log in for availability
    Each (1oz)
    Each (1oz) - $35.11
    $35.11
    Each (1oz)
       $35.11
    Supplier Number: 16909-01
  • 100504-868
    Immersion Oil Type NVH
    Immersion Oil Type NVH
    120 mL
    Log in for availability
    Log in for availability
    Each (120ml)
    Each (120ml) - $67.19
    $67.19
    Each (120ml)
       $67.19
    Supplier Number: 16909
  • 102090-988
    Immersion Oil Type OVH
    Immersion Oil Type OVH
    1 oz.
    Log in for availability
    Log in for availability
    Each
    Each - $35.95
    $35.95
    Each
       $35.95
    Supplier Number: 16918
  • 102092-090
    Immersion Oil Type OVH
    Immersion Oil Type OVH
    4 oz.
    Log in for availability
    Log in for availability
    Each
    Each - $34.44
    $34.44
    Each
       $34.44
    Supplier Number: 16918-01
  • 102092-092
    Immersion Oil Type OVH
    Immersion Oil Type OVH
    16 oz.
    Log in for availability
    Log in for availability
    Each
    Each - $194.33
    $194.33
    Each
       $194.33
    Supplier Number: 16918-16
  • 100496-520
    Immersion Oil Type 37
    Immersion Oil Type 37
    1 oz.
    Log in for availability
    Log in for availability
    Each (1oz)
    Each (1oz) - $33.60
    $33.60
    Each (1oz)
       $33.60
    Supplier Number: 16914-01
  • 100501-600
    Immersion Oil Type 37
    Immersion Oil Type 37
    4 oz.
    Log in for availability
    Log in for availability
    Each
    Each - $77.54
    $77.54
    Each
       $77.54
    Supplier Number: 16909-04