ContactUsLinkComponent
H.S.E. Test Slide for Phase Contrast Microscopy, Electron Microscopy Sciences
  100499-298
Minority or Woman-Owned Business Enterprise
undefined
H.S.E. Test Slide for Phase Contrast Microscopy, Electron Microscopy Sciences
  100499-298
 :  68038-84H
Minority or Woman-Owned Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Species:
  • Description:
    Hse test slides (s84) for calibration in asbestos analysis
  • Environmentally Preferable:
  • Cat. No.:
    100499-298
  • Supplier no.:
    68038-84H
Epoxy replica of a master slide. The replicas are mounted on microscope slides of 1.2 mm thickness with cover glass of 0.17 mm thickness.
This information is transferred to the accompanying table, which indicates the maximum phase change passing through test objects to test slide. A satisfactory system will detect block 5.
 : This test slide is made in the UK and is under license certification from the National Physical Laboratory.
 : Full details are supplied with the slide.