ContactUsLinkComponent
Graphite Conductive Adhesive 154, Electron Microscopy Sciences
Graphite Conductive Adhesive 154, Electron Microscopy Sciences
Catalog # 100496-462
Minority or Woman-Owned Business Enterprise
CAS Number:  
undefined
Graphite Conductive Adhesive 154, Electron Microscopy Sciences
Catalog # 100496-462
Supplier Number:  12691-30
Minority or Woman-Owned Business Enterprise
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Graphite Conductive Adhesive 154
  • Size:
    30 g
  • Cat. No.:
    100496-462
  • Supplier no.:
    12691-30
Specifications
About this item
Adhesive 154 is an easy-to-apply resistance coating designed to provide high lubricity, conductivity, and excellent release properties to many non-conductive substrates, including most plastics.
  • Easy-to-apply
  • High lubricity
  • Excellent release properties
Adhesive is made from a dispersion of colloidal graphite in an isopropanol base which quickly air dries and forms a uniform thin film adherent layer. Air dries to touch in 5 minutes and is ready for use in 30 minutes. After air drying, bake for 5 minutes at 167°F (75°C) to achieve optimum coating qualities in a shorter curing cycle.