ContactUsLinkComponent
Gold Shadowed Latex, Electron Microscopy Sciences
Gold Shadowed Latex, Electron Microscopy Sciences
  100496-368
Minority or Woman-Owned Business Enterprise
undefined
Gold Shadowed Latex, Electron Microscopy Sciences
  100496-368
 :  80040
Minority or Woman-Owned Business Enterprise
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    Gold Shadowed Latex
  • Cat. No.:
    100496-368
  • Supplier no.:
    80040
These latex particles have a diameter of 0.204 µm and are shadowed with a coating of gold. Measure the finest migrated gold particles in the shadow to determine the resolution of the STEM. Gold shadowed latex on formvar/carbon film, mounted on a copper grid, 3.05 mm diameter.