Glennel Lapping Solutions, Electron Microscopy Sciences
About this item
Penetrating formula designed to enhance cutting action and leaves a superior finish.
- Formula C is made especially for oil soluble compounds
Glenoil Lube - Lapping vehicle is the ideal lap oil for all Glennel® UB Compounds and Diamond Sprays. It assures continued separation of diamond particles and constant removal of lap debris.
Formula B is a universal lapping vehicle, compatible with both water and oil soluble compounds. It is ideal in an environment where materials change constantly. This very versatile formulation allows you to change materials and compounds and still provides excellent lubricity and cutting action.
3 Options Available Below
Product Details & Documents
Penetrating formula designed to enhance cutting action and leaves a superior finish.
- Formula C is made especially for oil soluble compounds
Glenoil Lube - Lapping vehicle is the ideal lap oil for all Glennel® UB Compounds and Diamond Sprays. It assures continued separation of diamond particles and constant removal of lap debris.
Formula B is a universal lapping vehicle, compatible with both water and oil soluble compounds. It is ideal in an environment where materials change constantly. This very versatile formulation allows you to change materials and compounds and still provides excellent lubricity and cutting action.
Product Family Options
Product Information
- SizeAvailabilityPrice
- 100497-394Glenoil LubeSupplier Number: 50376-01
- 100497-396Formula C LubeSupplier Number: 50376-05
- 100497-398Formula B LubeSupplier Number: 50376-10