- Description:5-Slide Unit Glass Coplin Jar, Screw Cap
- Cat. No.:100500-232
- Supplier no.:70317
Staining Jar holds 5 single 1 x 3" slides vertically, or 10 slides back-to-back.
Slides stay within the jar so that it can be capped during staining. Complete with a screw cap and a metal foil liner.
Overall measurement: 4 3/8 x 2 3/4 x 2 3/8.