About this item
This container will permit rapid specimen handling and provides temperature information during microwave irradiation. A liquid crystal strip is affixed into the bottom of a 35 mm diameter petri dish and covered with a thin layer of EMbed resin. The temperature range is 35°C-60°C. This container is ideal for tissue fixing by microwaves. The two active temperature windows are 45°C and 50°C.