Orders LinkContactUsLinkComponent
Fine Mesh Grids, Electron Microscopy Sciences
Fine Mesh Grids, Electron Microscopy Sciences
Fine Mesh Grids, Electron Microscopy Sciences
Certificates
Fine Mesh Grids, Electron Microscopy Sciences
Fine Mesh Grids, Electron Microscopy Sciences
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These fine mesh grids are carefully checked.
They are suitable for the low magnification range of a TEM.
Ordering information: Available as a sandwich in a folding 3.05 mm mesh grid, or in a square mesh 25 mm.
Product Family Options
Product Information
  • Material
    Type
    Availability
    Price
  • 100494-990
    Mesh Grid
    Mesh Grid
    Copper
    1000
    Log in for availability
    Log in for availability
    Each
    Each - $78.88
    $78.88
    Each
       $78.88
    Supplier Number: 79525-01
  • 100494-992
    Mesh Grid
    Mesh Grid
    Copper
    2000
    Log in for availability
    Log in for availability
    Each
    Each - $83.19
    $83.19
    Each
       $83.19
    Supplier Number: 79525-02
  • 100494-996
    Mesh Grid, Square, 25×25 mm
    Mesh Grid, Square, 25×25 mm
    Nickel
    2000
    Log in for availability
    Log in for availability
    Each
    Each - $344.67
    $344.67
    Each
       $344.67
    Supplier Number: 79526-20