Orders LinkContactUsLinkComponent
Film Casting Device, Electron Microscopy Sciences
Film Casting Device, Electron Microscopy Sciences
Certificates
Film Casting Device, Electron Microscopy Sciences
Film Casting Device, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
An all glass apparatus that casts uniformly thin films of parlodion, formvar, or butvar directly onto 1 x 3 microscope slides.
  • Film casting solution can be used repeatedly
  • Unit requires 100 mL of film casting solution to start
A built-in fine-pressure-release valve helps control the speed of drainage. The thickness of the film is controlled by the concentration of the film solution and the rate of the drainage.
Product Family Options
Product Information
  • Availability
    Price
  • 100493-270
    Film Casting Device Set
    Film Casting Device Set
    Log in for availability
    Log in for availability
    Each
    Each - $1,462.73
    $1,462.73
    Each
       $1,462.73
    Supplier Number: 71305-01
  • 100493-272
    500 mL Flask Replacement
    500 mL Flask Replacement
    Discontinued
    Supplier Number: -
  • 100493-274
    Film Casting Cylinder Replacement
    Film Casting Cylinder Replacement
    Log in for availability
    Log in for availability
    Each
    Each - $530.19
    $530.19
    Each
       $530.19
    Supplier Number: 71305-06