About this item
Prepared with subnanometer gold particles, these particles have far less influence on the adsorbed antibodies or detecting molecules, and consequently behave as though they are uncoupled. Steric hindrance is decreased compared to conventional.
Aurion Ultra Small Immunogold Reagents are prepared with subnanometer gold particles. These particles have far less influence on the adsorbed antibodies or detecting molecules, and consequently the conjugates behave as though they are uncoupled. In conjunction with the highly efficient and easy-to-use R-Gent SE-LM and SE-EM silver enhancement reagents, the Ultra Small Immunogold Reagents are the best choice for any application.Reduction of the gold particle size provides Ultra Small Immunogold Reagents with fundamentally different characteristics when compared with conjugates built around larger particles. While the Conventional Immunogold Reagents can be thought of as particles coated with proteins, Ultra Small Immunogold Reagents are proteins coated with one or more gold particles. With this structure, both the overall size of the conjugates, as well as steric hindrance are decreased.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Gold, >1 nm (ultra-small)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 100487-766
- 100487-768
- Antigen Symbol:
- IgG
- IgG
- Antigen Name:
- Immunoglobulin G
- Immunoglobulin G
- Antibody Type:
- Secondary
- Secondary
- Clonality:
- Polyclonal
- Polyclonal
- Reactivity:
- Mouse
- Mouse
- Host:
- Goat
- Goat
- Isotype:
- IgG
- IgG