About this item
EMbed 812 has the same characteristics as Epon 812, physically, and provides the same results in preservation, handling, curing and sectioning.
- Specific Gravity: 1.08-1.10
EMbed 812 may cause etching on selected plastics. All formulas for mixing and using Epon 812 can be applied to EMbed 812, e.g. Luft's procedure, Mollenhauer's and EPON-ARALDITE mixtures.