About this item
Rotary vacuum pump; Double stage. Ultra Grade 19 oil is recommended for use with the RV5. Weight 43 lbs. (19.6 kg).
- 4.1 cfm (117L/min) displacement
- Ultimate vacuum (without gas ballast) 1.5x10-3 torr
- Max. inlet pressure for water vapor: 38 torr
- Max. water vapor pumping rate: 0.48 lb/h
- Motor: 1/2 hp
- Oil capacity Max/Min 0.7L/0.42L