About this item
A water soluble embedding medium for EM based on an aliphatic polyepoxide.
Fluka Durcupan embedding agent opens new roads in the observation of enzymatic digestion processes and for carrying out histochemical investigations in the submicroscopic range. It enables formalin and other aqueous fixatives to be used for electron microscopy and the dehydration of the tissue in alcohol or acetone to be dispensed with, so that excessive changes in the ultrastructure of the cells are obviated.