Dissecting Kit, 8 pieces, Stainless Steel, Electron Microscopy Sciences
Catalog # 100491-300
Supplier: Electron Microscopy Sciences
Minority or Woman-Owned Business Enterprise
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:8-Piece Stainless Steel Dissecting Kit
- Cat. No.:100491-300
- Supplier no.:72950
Specifications
About this item
This dissection kit includes the following:
- 1 4½" Dissecting forceps with curved and serrated tips
- 1 Curved dissecting needle
- 1 Glass pipette with rubber bulb
- 1 4½" Dissecting forceps straight and serrated tips
- 1 Dissecting scalpel, 6" long, 1½" cuting edge.
- 1 4½" Dissecting scissors, sharp, straight points
- 1 Plastic ruler 6", 1/16" div., millimeter and centimeter