COVAMAT® Lining, Electron Microscopy Sciences
Versatile clear lining material: clear polyethylene ribbed matting can be used for lining laboratory bench top, shelves, trays, and more.
It is easily trimmed and also works well under drums or carboys. 0.25 mm (0.010") width 0.7 mm (0.030") thick ribs, 76 cm (30") wide, 15.2 m (50') and 4.6 m (15') roll. 1 roll/box.
Versatile clear lining material: clear polyethylene ribbed matting can be used for lining laboratory bench top, shelves, trays, and more.
It is easily trimmed and also works well under drums or carboys. 0.25 mm (0.010") width 0.7 mm (0.030") thick ribs, 76 cm (30") wide, 15.2 m (50') and 4.6 m (15') roll. 1 roll/box.
Product Family Options
- DescriptionCapacity
- Covamat Lining, 15'15 ft-: -
- : 72615-50