About this item
Histology control slides have patient and control tissue samples.
- Positive identification of control and patient samples
- Proof of correct staining procedures
- Utmost adhesion of samples to slide throughout staining procedure
- Easy and permanent identification of slides
Available in two configurations: Style 329+ with ¾" of frosted area and Style 334+ with 1" frosted area.