ContactUsLinkComponent
Butler Block Trimmer, Electron Microscopy Sciences
Butler Block Trimmer, Electron Microscopy Sciences
Catalog # 100492-872
Minority or Woman-Owned Business Enterprise
undefined
Butler Block Trimmer, Electron Microscopy Sciences
Catalog # 100492-872
Supplier Number:  69945-01
Minority or Woman-Owned Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Butler Block Trimmer
  • Cat. No.:
    100492-872
  • Supplier no.:
    69945-01
Specifications
About this item
A precision made device to aid in hand trimming resin tissue blocks to the desired shape for ultramicrotomy.
  • Used with any stereomicroscope with epi-illumination
The knurled knife guide swivels to help the trimming blade to cut the sides of the block.
Ordering information: The trimmer is supplied with a standard chuck to accommodate the block size 00.