Atomizer Rubber Bulb, Electron Microscopy Sciences
A natural rubber bulb with 4" tubing on one end. Used for dusting surfaces which should not be wiped.
A two-way valve is inserted into one end to prevent suck back and a rubber nozzle prevents scratches on the surface that have to be cleaned. This device also reduces the need for freon dusters.
A natural rubber bulb with 4" tubing on one end. Used for dusting surfaces which should not be wiped.
A two-way valve is inserted into one end to prevent suck back and a rubber nozzle prevents scratches on the surface that have to be cleaned. This device also reduces the need for freon dusters.
Product Family Options
- Description
- : 60806
- : 60804