Invoil Oil BW
- Consists of esters or specially selected hydrocarbon fractions
- High purity, very low pressure, very stable
- Pressures lower than 0.013 – 0.009 bar obtainable
- No need for cold trap
- Good pumping speed
- No corrosive deposits or vapors
Invoil Oil C
Obtains the highest vacuum pressure (0.013 – 0.009 bar) of all of the oils without the need of a cold trap.
Invoil Oil BW: A water-white oil with a higher pumping speed than oil B, although all other characteristics are similiar to B. Because it has an extremely high purity it is suitable for use in the evacuation of mass spectrometers.
Invoil Oil C: Obtains the highest vacuum pressure (0.013 – 0.009 bar) of all of the oils without the need of a cold trap.