- Description:Grease, Type T
- Size:25 g
- Cat. No.:100504-356
- Supplier no.:60704
Type T is designed for lubricating glass taps and for general purposes. Has a melting point of 125°C and has an estimated vapor pressure @ 20°C of 5x10-9 torr. Although type T is useful at higher temperatures, it can also be used at ambient temperatures. Temperature ranges 0-120°C.