Order Entry
Puerto Rico
ContactUsLinkComponent
 
Hard Plus Resin, Electron Microscopy Science
  100503-436
Minority or Woman-Owned Business Enterprise
 :  
Hard Plus Resin, Electron Microscopy Science
  100503-436
 :  14115
Minority or Woman-Owned Business Enterprise
 :  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Description:
    Hard Plus Resin 812 Kit
  • Density:
    0.98 g/cm³ (8.178 lbs/gal)
  • Boiling Point:
    180 °C (356 °F)
  • Cat. No.:
    100503-436
  • Supplier no.:
    14115

 

 

Hard-Plus Resin-812 is supplied only as a premix kit

A newly formulated resin that enables the processing of specimens that have a tendency to wrinkle and be problematic.

In general, Hard Plus Resin-812 and its WPE numbers (weight per epoxide) do not vary much from batch to batch and it also has good stability under the electron beam and it stains well with heavy metals. Furthermore, the viscosity of the Hard-Plus Resin-812 is lower than the original Epon-812, making infiltration of the tissue more complete. For processing and embedding, it can be used exactly the same way as Epon-812, but normally, it is not necessary to use intermediate solvents, such as propylene oxide, hence, infiltration can immediately follow with absolute alcohol. Polymerization is normally at 60°C for 24 – 48 hours. Longer times or higher temperatures (70°C or more) will give a harder block.

For larger samples (>1x1mm3) the use of a intermediate solvent is recommended.

Hard-Plus Resin-812 is supplied only as a premix kit. All components are pre-measured, and you have to do is combine them in the right order and mix thoroughly. Each kit contains five separate mini-kits, making approximately 100g each.

Kit consists of: 5x50g Hard-Plus Resin-812, 5x50g Hardener Hard-Plus, 5x 2.5 ml Accelerator, Hard-Plus Resin-812 Kit