About this item
CryoStor® cryopreservation freeze media products have been designed to mitigate temperature-induced molecular cell stress responses during freezing and thawing
Across a broad spectrum of cell types, CryoStor products have proven much more effective in reducing post-preservation necrosis and apoptosis as compared to commercial and home-brew isotonic and extracellular formulations. This enables greatly improved post-thaw cell yield, viability, and recovery. CryoStor products meet USP (71) Sterility and USP (85) Endotoxin testing standards, and are manufactured under cGMP.
CryoStor is offered in 100 mL bottles and pre-formulated with DMSO in final concentrations of 2%, 5%, and 10%.
CryoStor proprietary freeze media products are intended for cryopreservation of biologics at -80 to -196°C and are based on the novel HypoThermosol formula. All CryoStor products are pre-formulated with USP grade DMSO, a permeant solute cryoprotective agent which helps mitigate damage from the formation of intracellular ice.