About this item
The Bulldog clamp is a useful instrument for stopping blood flow to a particular organ of interest
One of the most common applications where a bulldog clamp can be used is during coronary surgery where blood flow must be stopped. In general the bulldog clamp, with its many angled varieties and surgical applications, is an essential tool during surgery where blood flow to an organ must be stopped or controlled
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Catalog No:
- 100188-818
- 100188-902
- Jaw Length:
- 8 mm
- 8 mm
- Jaw Width:
- 1.2 mm
- 1.2 mm
- Pressure:
- 80g
- 80g