About this item
This antibody reacts with an antigen expressed by normal and neoplastic T cells and myeloid lineage cells. It strongly stains T cells in the interfollicular areas of normal lymphoid tissues.
Type:
Antigen: CD43 (T Cell)
Clonality: Monoclonal
Clone: DF-T1
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 99990-192
- 95024-348
- 99990-908
- Antigen Symbol:
- CD43 (T Cell)
- CD43 (T Cell)
- CD43 (T Cell)
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- DF-T1
- DF-T1
- DF-T1
- Host:
- Mouse
- Mouse
- Mouse
- Concentration:
- Concentrated
- Concentrated
- Ready to use