ContactUsLinkComponent
SP Bel-Art® Tapered Plug Sampler, Bel-Art Products, a part of SP
SP Bel-Art® Tapered Plug Sampler, Bel-Art Products, a part of SP
Catalog # 70680-276
Supplier:  VWR International
undefined
SP Bel-Art® Tapered Plug Sampler, Bel-Art Products, a part of SP
Catalog # 70680-276
Supplier:  VWR International
Specifications
  • Length:
    19.1 cm (7½”)
  • Cat. No.:
    70680-276
  • Supplier no.:
    378260000
Specifications
About this item
A smooth, tapered, stainless sampler for extracting a “plug” of material from bags or blocks of cheese, grease, soap, clay, etc.
  • Stainless steel construction
  • Can easily cut through a bag or box
  • Smooth, tapered design
The sharp end of the satin finish sampler will cut through a bag or box.
Twisting the handle one-half turn will cut out a plug of material 15.2 cm (6”) long tapering from 1.7 cm (¹¹/₁₆”) to 1.3 cm (½”). “T” handle is 7 cm (2¾”).