SP Bel-Art Stainless Steel Coated Lab Spoon and Spatula, Bel-Art Products, a part of SP
70690-180
- Description:Double-ended, coated stainless steel, lab spoon/spatula
- Length:22.9 cm (9")
- Cat. No.:70690-180
- Supplier no.:367110000
This stainless steel sampling spoon and spatula has ends coated with brown, Fluo-Kem® Teflon® FEP that complies with FDA regulations governing direct food contact and provides an anti-stick, corrosion-resistant surface.
- FEP-coated stainless steel construction
- Coating provides anti-stick, corrosion-resistant surface
- Two ends are spatula and spoon
The 4 mm (⁵⁄₃₂") Ø rod is flattened at one end to give an 8×51 mm (⁵⁄₁₆×2") spatula. The opposite end is a 13×25 mm (½×1") spoon.