
3-Methylflavone-8-carboxylic acid for synthesis, Sigma-Aldrich®
Catalog # EM8.41878.0005
Supplier: MilliporeSigma
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle, pre-packed
- Size:5 g
- Cat. No.:EM8.41878.0005
- Supplier no.:8.41878.0005
Specifications
Specification Test Results
- Melting Point:235 - 240 °C
- Molar Mass:280.28 g/mol
Specification Test Results
About this item
:
:280.28 g/mol
Boiling Pt:211 °C (1013 hPa)
Density:1.408 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:259-193-7
:GHS07
Storage Temperature:Ambient
CAS Number:3468-01-7
Formula:C₁₇H₁₂O₄
MDL Number:MFCD00040998