Order Entry
Puerto Rico
ContactUsLinkComponent
 
Corrosive
2,3,5-Trichlorobenzaldehyde for synthesis, Sigma-Aldrich®
Catalog # EM8.41271.0001
Supplier:  MilliporeSigma
CAS Number:  
Corrosive
2,3,5-Trichlorobenzaldehyde for synthesis, Sigma-Aldrich®
Catalog # EM8.41271.0001
Supplier:  MilliporeSigma
Supplier Number:  8.41271.0001
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 g
  • Cat. No.:
    EM8.41271.0001
  • Supplier no.:
    8.41271.0001

Specifications

Specification Test Results

  • Melting Point:
    75 - 76 °C
  • Molar Mass:
    209.45 g/mol

Specification Test Results

About this item

 :
 :209.46 g/mol
Boiling Pt:359 °C (1013 hPa)
Melting Pt:75…76 °C
Storage Temperature:Ambient
ADR:8,II
 :M-Clarity™ quality level = MQ200
UN:1759
 :GHS05
CAS Number:56961-75-2
Formula:C₇H₃Cl₃O
MDL Number:MFCD00010860