Order Entry
Puerto Rico
ContactUsLinkComponent
 
Flammable
4,4-Dimethyl-2-oxazoline for synthesis, Sigma-Aldrich®
Catalog # EM8.18107.0005
Supplier:  MilliporeSigma
CAS Number:  
Flammable
4,4-Dimethyl-2-oxazoline for synthesis, Sigma-Aldrich®
Catalog # EM8.18107.0005
Supplier:  MilliporeSigma
Supplier Number:  8.18107.0005
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    5 ml
  • Cat. No.:
    EM8.18107.0005
  • Supplier no.:
    8.18107.0005

Specifications

Specification Test Results

  • Solubility:
    (20 °C) (decomposition)
  • Molar Mass:
    99.13 g/mol
  • Boiling Point:
    99 - 100 °C (1013 hPa)
  • Flash Point:
    1 °C
  • Density:
    0.94 g/cm3 (20 °C)

Specification Test Results

About this item

 :
 :99.13 g/mol
ADR:3,II
 :M-Clarity™ quality level = MQ200
UN:1993
 :GHS02
CAS Number:30093-99-3
Formula:C₅H₉NO
MDL Number:MFCD00014507