Order Entry
Puerto Rico
ContactUsLinkComponent
2-Chloropropyldimethylammonium chloride for synthesis, Sigma-Aldrich®
Irritant
2-Chloropropyldimethylammonium chloride for synthesis, Sigma-Aldrich®
Catalog # EM8.03240.0250
Supplier:  MilliporeSigma
CAS Number:  
Irritant
2-Chloropropyldimethylammonium chloride for synthesis, Sigma-Aldrich®
Catalog # EM8.03240.0250
Supplier:  MilliporeSigma
Supplier Number:  8.03240.0250
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    250 g
  • Cat. No.:
    EM8.03240.0250
  • Supplier no.:
    8.03240.0250

Specifications

Specification Test Results

  • Solubility:
    2000 g/l (20 °C)
  • Melting Point:
    188 - 191 °C
  • Molar Mass:
    158.07 g/mol
  • Bulk Density:
    350 kg/m3
  • Flash Point:
    220 °C
  • pH:
    5 - 6 (500 g/l, H2O, 20 °C)

Specification Test Results

About this item

 :
 :158.07 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:224-971-7
 :GHS07
CAS Number:4584-49-0
Formula:C₅H₁₃Cl₂N
MDL Number:MFCD00012534