Ammonium Standard Solution, CRM traceable to SRM from NIST 6,00 mg/L NH₄-N in H₂O
Catalog # EM1.25025.0100
Supplier: MilliporeSigma
CAS Number:
Specifications
- Concentration:6 ppm
- Pack type:Plastic Bottle
- Description:Ammonium standard solution
- Size:100 ml
- Cat. No.:EM1.25025.0100
- Contents:NH₄Cl in H₂O
- Supplier no.:1.25025.0100