Order Entry
Puerto Rico
ContactUsLinkComponent
 
Ammonium Standard Solution, CRM traceable to SRM from NIST 1,00 mg/L NH₄-N in H₂O
Catalog # EM1.25023.0100
Supplier:  MilliporeSigma
CAS Number:  
Ammonium Standard Solution, CRM traceable to SRM from NIST 1,00 mg/L NH₄-N in H₂O
Catalog # EM1.25023.0100
Supplier:  MilliporeSigma
Supplier Number:  1.25023.0100
CAS Number:  

Specifications

  • Concentration:
    1 ppm
  • Pack type:
    Plastic Bottle
  • Description:
    Ammonium standard solution
  • Size:
    100 ml
  • Cat. No.:
    EM1.25023.0100
  • Contents:
    NH₄Cl in H₂O
  • Supplier no.:
    1.25023.0100

Specifications

About this item