ContactUsLinkComponent
Chromate Test (Chromium VI) Method: photometric 0.010 - 3.00 mg/L Cr 0.02 - 6.69 mg/L CrO₄ Spectroquant®
Chromate Test (Chromium VI) Method: photometric 0.010 - 3.00 mg/L Cr 0.02 - 6.69 mg/L CrO₄ Spectroquant®
Catalog # EM1.14758.0001
Supplier:  MilliporeSigma
CAS Number:  
Chromate Test (Chromium VI) Method: photometric 0.010 - 3.00 mg/L Cr 0.02 - 6.69 mg/L CrO₄ Spectroquant®
Catalog # EM1.14758.0001
Supplier:  MilliporeSigma
Supplier Number:  1.14758.0001
CAS Number:  
Specifications
  • Pack type:
    Cardboard Box(es)
  • Size:
    1 pc
  • Cat. No.:
    EM1.14758.0001
  • Supplier no.:
    1.14758.0001