ContactUsLinkComponent
Formaldehyde Test Method: photometric 0.02 - 8.00 mg/l HCHO Spectroquant®
Formaldehyde Test Method: photometric 0.02 - 8.00 mg/l HCHO Spectroquant®
Catalog # EM1.14678.0001
Supplier:  MilliporeSigma
CAS Number:  
Formaldehyde Test Method: photometric 0.02 - 8.00 mg/l HCHO Spectroquant®
Catalog # EM1.14678.0001
Supplier:  MilliporeSigma
Supplier Number:  1.14678.0001
CAS Number:  
Specifications
  • Pack type:
    Cardboard Box(es)
  • Size:
    1 pc
  • Cat. No.:
    EM1.14678.0001
  • Supplier no.:
    1.14678.0001