Nickel Cell Test Method: Photometric 0.10 - 6.00 mg/L Ni Spectroquant®, MilliporeSigma
Catalog # EM1.14554.0001
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:EPS-Moulding
- Size:1 pc
- Cat. No.:EM1.14554.0001
- Supplier no.:1.14554.0001