Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Environmental Test Kits
Total Hardness Test Method: titrimetric with dropping bottle MColortest™
Total Hardness Test Method: titrimetric with dropping bottle MColortest™
Total Hardness Test Method: titrimetric with dropping bottle MColortest™
Catalog # EM1.11104.0001
Supplier:
MilliporeSigma
CAS Number:
SDS
Certificates
Expand image
Total Hardness Test Method: titrimetric with dropping bottle MColortest™
Catalog # EM1.11104.0001
Supplier:
MilliporeSigma
Supplier Number:
1.11104.0001
CAS Number:
SDS
Certificates
Specifications
Pack type:
Plastic Box(es)
Size:
1 pc
Cat. No.:
EM1.11104.0001
Supplier no.:
1.11104.0001
Specifications
About this item
Product Details
Documentation
Product Details
Documentation