ContactUsLinkComponent
Phosphate Test Strips, MQuant®
Catalog # EM1.10428.0001
Supplier:  MilliporeSigma
CAS Number:  
Phosphate Test Strips, MQuant®
Catalog # EM1.10428.0001
Supplier:  MilliporeSigma
Supplier Number:  1.10428.0001
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Pack type:
    Plastic Box(es)
  • Description:
    Phophate test strips
  • Size:
    1 pc
  • Cat. No.:
    EM1.10428.0001
  • Range:
    10 - 500 mg/L PO₄³⁻
  • Supplier no.:
    1.10428.0001
  • No. of tests:
    100
Specifications
About this item
MQuant™ test strips can be considered as a mobile laboratory on a few square millimeters of plastic strip. As a screening method they provide the user with a rapid overview of the situation, which helps in making the choice of what, and perhaps if, further analysis is required, thereby leading to potentially substantial savings in time and analytical resources. The strip is a biologically degradable polyester foil and the low reagent content, in the reaction zones, assists problem-free waste disposal.
  • Easy to handle: Just dip into sample, add reagent, wait and measure visually against colour scale
  • Immediate results within 2 minutes enables fast decisions
  • Portable: Can be used inside or outside the laboratory
The phosphate test is suitable for determining orthophosphate in wastewater, soil samples, fertilizers and food.
Gradation: 10 - 25 - 50 - 100 - 250 - 500 mg/l PO₄³⁻