Formaldehyde Test Strips, MQuant®
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
MQuant™ test strips can be considered as a mobile laboratory on a few square millimeters of plastic strip. As a screening method they provide the user with a rapid overview of the situation, which helps in making the choice of what, and perhaps if, further analysis is required, thereby leading to potentially substantial savings in time and analytical resources. The strip is a biologically degradable polyester foil and the low reagent content, in the reaction zones, assists problem-free waste disposal.
- Easy to handle: Just dip, wait and measure visually against colour scale
- Safe: No auxiliary reagents necessary
- Immediate results within 2 minutes enables fast decisions
- Portable: Can be used inside or outside the laboratory
Formaldehyde reacts with 4-amino-3-hydrazino5-mercapto-1,2,4-triazole to form a purple-red tetrazine. The formaldehyde concentration is measured semiquantitatively by visual comparison of the reaction zone of the test strip with the fields of a color scale.
Gradation: 10 - 20 - 40 - 60 - 100 mg/l HCHO
Applications: Disinfectant and rinsing solutions (e.g. laundries), aqueous solutions.
Documents
Certificates
Search by Lot NumberEMD Millipore Simply Rapid MQuant and MColorpHast Mobile Analysis System
Product Literature
ghs_1.10036
SDS
VWR GHS Compliance Letter (US)
Additional Product Details
Specifications
Specifications
- Chemical method:Triazole
- Pack type:Plastic Box(es)
- Description:Formaldehyde Test Strips
- Size:1pc
- Range:10 - 100 mg/L HCHO
- No. of tests:100
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



