Order Entry
Puerto Rico
ContactUsLinkComponent
Magnesium, foil, Width 3 mm, Thickness 0.15-0.30 mm, Supelco®
Magnesium, foil, Width 3 mm, Thickness 0.15-0.30 mm, Supelco®
Catalog # EM1.05812.0001
Supplier:  MilliporeSigma
CAS Number:  
Magnesium, foil, Width 3 mm, Thickness 0.15-0.30 mm, Supelco®
Catalog # EM1.05812.0001
Supplier:  MilliporeSigma
Supplier Number:  1.05812.0001
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Cardboard box
  • Size:
    1 piece
  • Cat. No.:
    EM1.05812.0001
  • Supplier no.:
    1.05812.0001

Specifications

Specification Test Results

  • Melting Point:
    651 °C
  • Molar Mass:
    24.31 g/mol
  • Boiling Point:
    1107 °C
  • Density:
    1.75 g/cm3 (20 °C)

Specification Test Results

About this item

 :
 :24.3 g/mol
UN:1869
ADR:4.1,III
 :GHS02
CAS Number:7439-95-4
Formula:Mg
MDL Number:MFCD00085308