Order Entry
Puerto Rico
ContactUsLinkComponent
 
Toxic
Health
Environment
Cadmium, granular, EMSURE® ∼ 3-6 mm for analysis, Supelco®
Catalog # EM1.02004.0250
Supplier:  MilliporeSigma
CAS Number:  
Toxic
Health
Environment
Cadmium, granular, EMSURE® ∼ 3-6 mm for analysis, Supelco®
Catalog # EM1.02004.0250
Supplier:  MilliporeSigma
Supplier Number:  1.02004.0250
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Metal can with PE liner
  • Size:
    250 g
  • Cat. No.:
    EM1.02004.0250
  • Supplier no.:
    1.02004.0250

Specifications

Specification Test Results

  • Cu (Copper):
    ≤ 0.001 %
  • Fe (Iron):
    ≤ 0.001 %
  • Hg (Mercury):
    ≤ 0.0005 %
  • Pb (Lead):
    ≤ 0.005 %
  • Zn (Zinc):
    ≤ 0.001 %

Specification Test Results

About this item

 :
 :112.41 g/mol
Merck Index:12,01649
Melting Pt:321 °C
Density:8.6 g/cm3 (22 °C)
Storage Temperature:Ambient
EINECS:231-152-8
UN:3288
ADR:6.1,III
 :GHS06
 :GHS08
 :GHS09
CAS Number:7440-43-9
Formula:Cd
MDL Number:MFCD00010914