Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Environmental Test Kits
Spectroquant® Phenol Test Method: photometric 0.002 - 5.00 mg/L
Spectroquant® Phenol Test Method: photometric 0.002 - 5.00 mg/L
Spectroquant® Phenol Test Method: photometric 0.002 - 5.00 mg/L
Catalog # EM1.00856.0001
Supplier:
MilliporeSigma
CAS Number:
SDS
Expand image
Spectroquant® Phenol Test Method: photometric 0.002 - 5.00 mg/L
Catalog # EM1.00856.0001
Supplier:
MilliporeSigma
Supplier Number:
1.00856.0001
CAS Number:
SDS
Specifications
Pack type:
Cardboard Box(es)
Size:
1 pc
Cat. No.:
EM1.00856.0001
Supplier no.:
1.00856.0001
Specifications
About this item
Product Details
Documentation
Product Details
Documentation