ContactUsLinkComponent
Chlorine Reagent Cl₂-1 (liquid) for Chlorine Test (DPD) 0.010 - 6.00 mg/L Cl₂ free chlorine: Cl₂-1 and Cl₂-2 total chlorine: Cl₂-1, Cl₂-2, and Cl₂-3 Spectroquant®
Chlorine Reagent Cl₂-1 (liquid) for Chlorine Test (DPD) 0.010 - 6.00 mg/L Cl₂ free chlorine: Cl₂-1 and Cl₂-2 total chlorine: Cl₂-1, Cl₂-2, and Cl₂-3 Spectroquant®
Catalog # EM1.00086.0001
Supplier:  MilliporeSigma
CAS Number:  
Chlorine Reagent Cl₂-1 (liquid) for Chlorine Test (DPD) 0.010 - 6.00 mg/L Cl₂ free chlorine: Cl₂-1 and Cl₂-2 total chlorine: Cl₂-1, Cl₂-2, and Cl₂-3 Spectroquant®
Catalog # EM1.00086.0001
Supplier:  MilliporeSigma
Supplier Number:  1.00086.0001
CAS Number:  
Specifications
  • Pack type:
    Plastic Bottle
  • Size:
    1 pc
  • Cat. No.:
    EM1.00086.0001
  • Supplier no.:
    1.00086.0001