Separatory Globe Funnel, Ace Glass
89085-040
Product available on GSA Advantage®
Small Business Enterprise
- Capacity:125 ml
- Description:Separatory Globe Funnel
- Cat. No.:89085-040
- Stem length:175 mm
- Supplier no.:7221-09
This Globe-Style Separatory Funnel features a 175 mm stem below the PTFE stopcock and is supplied with a flask length top stopper.