Order Entry
Puerto Rico
ContactUsLinkComponent
 
Bordetella pertussis FHA, Enzo Life Sciences
Catalog # 10033-538
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Bordetella pertussis FHA, Enzo Life Sciences
Catalog # 10033-538
Supplier:  Enzo Life Sciences
Supplier Number:  630-123-0100
CAS Number:  

Specifications

  • Size:
    100 µg
  • Purity:
    >98%
  • Formulation:
    Lyophilized. Contains 0.05M sodium phosphate and 0.5M sodium chloride.
  • Cat. No.:
    10033-538
  • Supplier no.:
    630-123-0100

Specifications

About this item

From Bordetella pertussis, strain NCTC 10739.