C. difficile Toxin B, Enzo Life Sciences
50 µg
Supplier: Enzo Life Sciences
Supplier #: BML-G150-0050
<i>C. difficile</i> Toxin B, Enzo Life Sciences
Catalog # 10033-552
Supplier: Enzo Life Sciences
CAS Number:
Jump to:
Product Details & Documents
From Clostridium difficile, strain VPI10463.
Documents
Specifications
Specifications
- Size:50 µg
- Purity Detail:Purified protein.
- Cat. No.:10033-552
- Biological Activity:4 pg/ml by verocytotoxicity assay (minimum conconcentration at which cell rounding is visible)
- Formulation:Lyophilized. Contains 0.05M HEPES, 0.15M sodium chloride and 5% sucrose.
- Purity:>95% (SDS-PAGE)



