Bordetella pertussis toxin, Enzo Life Sciences
Catalog # 10033-540
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Size:50 µg
- Purity Detail:Purified protein.
- Biological Activity:0.03 ng/ml by CHO cell assay (minimum concentration at which cell rounding is visible)
- Purity:>98%
- Formulation:Lyophilized. Contains 0.05M sodium phosphate and 0.5M sodium chloride.
- Cat. No.:10033-540
- Supplier no.:BML-G101-0050
Specifications
About this item
High purity toxin with minimal lot-to-lot variation.
From Bordetella pertussis, strain NCTC 10739.
Native protein.